siRNA Details
| Gene Symbol: 4930519N06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK015846.1 | - | 1 | 667 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY715391.1 | - | 1 | 665 | Chr. 11 - 34093244 - 34094126 [-] on Build GRCm38 | |

4930519N06Rik
Genomic Map