siRNA Details
Gene Symbol: 4930519N06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK015846.1 | - | 1 | 348 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY715391.1 | - | 1 | 346 | Chr. 11 - 34093244 - 34094126 [-] on Build GRCm38 |
