siRNA Details
| Gene Symbol: 4931419H13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040593.1 | 8 | 946 | |||
| GenBank mRNA | AK016464.1 | - | 8 | 946 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4931419H13Rik| Gene Symbol: 4931419H13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040593.1 | 8 | 946 | |||
| GenBank mRNA | AK016464.1 | - | 8 | 946 | ||