siRNA Details
| Gene Symbol: 4931419H13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040593.1 | 6 | 763 | |||
| GenBank mRNA | AK016464.1 | - | 6 | 763 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY716138.1 | - | 6 | 756 | Chr. 3 - 55055246 - 55083975 [+] on Build GRCm38 | |

4931419H13Rik
Genomic Map