siRNA Details
| Gene Symbol: 4931418L13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK016462.1 | - | 1 | 78 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY716135.1 | - | 1 | 78 | Chr. 11 - 101347973 - 101348573 [+] on Build GRCm38 | |

4931418L13Rik
Genomic Map