siRNA Details
| Gene Symbol: Rbm3os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033561.1 | 2 | 318 | |||
| GenBank mRNA | AK013472.1 | - | 2 | 318 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BE655728.1 | - | 2 | 199 | Chr. X - 8145546 - 8147713 [+] on Build GRCm38 | |

Rbm3os
Genomic Map