siRNA Details
| Gene Symbol: 2210009P08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK008691.1 | - | 1 | 61 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY708583.1 | - | 1 | 61 | Chr. 16 - 97466484 - 97466948 [-] on Build GRCm38 | |

2210009P08Rik
Genomic Map