siRNA Details
| Gene Symbol: Gm30094 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC053977.1 | - | 4 | 665 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm30094| Gene Symbol: Gm30094 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC053977.1 | - | 4 | 665 | ||