siRNA Details
| Gene Symbol: Gm30094 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC053977.1 | - | 1, 2 | 107 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BE632065.1 | - | 1, 2 | 95 | Chr. 18 - 65863548 - 65894755 [-] on Build GRCm38 | |

Gm30094
Genomic Map