siRNA Details
| Gene Symbol: 1700030L20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040592.1 | 2, 3 | 502 | |||
| GenBank mRNA | AK006559.1 | - | 2, 3 | 503 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700030L20Rik| Gene Symbol: 1700030L20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040592.1 | 2, 3 | 502 | |||
| GenBank mRNA | AK006559.1 | - | 2, 3 | 503 | ||