siRNA Details
| Gene Symbol: A230107N01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK053270.1 | - | 2 | 1952 | ||
| BB537031.2 | - | 1 | 295 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

A230107N01Rik| Gene Symbol: A230107N01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK053270.1 | - | 2 | 1952 | ||
| BB537031.2 | - | 1 | 295 | |||