siRNA Details
| Gene Symbol: A230107N01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK039207.1 | - | 1 | 378 | ||
| AK053270.1 | - | 2 | 207 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB606329.2 | - | 2 | 207 | Chr. 13 - 84221301 - 84222175 [-] on Build GRCm38 | |

A230107N01Rik
Genomic Map