siRNA Details
| Gene Symbol: Pgls | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK080749.1 | - | 1398 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB282517.2 | - | 1 | 504 | Chr. 8 - 71590897 - 71591573 [-] on Build GRCm38 | |

Pgls
Genomic Map