siRNA Details
| Gene Symbol: AW549542 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK035112.1 | - | 26 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB627380.1 | - | 1, 2 | 26 | Chr. 5 - 119575154 - 119627305 [-] on Build GRCm38 | |

AW549542
Genomic Map