siRNA Details
| Gene Symbol: AW549542 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045702.1 | 2 | 936 | |||
| GenBank mRNA | AK035112.1 | - | 958 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

AW549542| Gene Symbol: AW549542 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045702.1 | 2 | 936 | |||
| GenBank mRNA | AK035112.1 | - | 958 | |||