siRNA Details
| Gene Symbol: AW549542 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045702.1 | 2 | 381 | |||
| GenBank mRNA | AK035112.1 | - | 403 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB627380.1 | - | 3 | 403 | Chr. 5 - 119575154 - 119627305 [-] on Build GRCm38 | |

AW549542
Genomic Map