siRNA Details
| Gene Symbol: Gm13490 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040639.1 | 5 | 1245 | |||
| GenBank mRNA | AK028256.1 | - | 5 | 1242 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm13490| Gene Symbol: Gm13490 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040639.1 | 5 | 1245 | |||
| GenBank mRNA | AK028256.1 | - | 5 | 1242 | ||