siRNA Details
| Gene Symbol: E230032D23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK037811.1 | - | 1 | 626 | ||
| AK154738.1 | - | 1 | 371 | |||
| BB160234.2 | - | 1 | 572 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

E230032D23Rik| Gene Symbol: E230032D23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK037811.1 | - | 1 | 626 | ||
| AK154738.1 | - | 1 | 371 | |||
| BB160234.2 | - | 1 | 572 | |||