siRNA Details
| Gene Symbol: AU022793 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045719.1 | 2, 3 | 353 | |||
| GenBank mRNA | AK155489.1 | - | 2, 3 | 356 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY749221.1 | - | 2, 3 | 356 | Chr. 15 - 39962649 - 39966257 [+] on Build GRCm38 | |

AU022793
Genomic Map