siRNA Details
| Gene Symbol: Znrf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK078666.1 | - | 991 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | EH109114.1 | - | 1 | 40 | Chr. 6 - 54890121 - 54890678 [+] on Build GRCm38 | |

Znrf2
Genomic Map