siRNA Details
| Gene Symbol: A630023A22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK041587.1 | - | 5 | 1189 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

A630023A22Rik| Gene Symbol: A630023A22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK041587.1 | - | 5 | 1189 | ||