siRNA Details
| Gene Symbol: LOC100505025 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_105062.1 | 1 | 142 | |||
| GenBank mRNA | AK133139.1 | - | 1 | 143 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100505025| Gene Symbol: LOC100505025 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_105062.1 | 1 | 142 | |||
| GenBank mRNA | AK133139.1 | - | 1 | 143 | ||