siRNA Details
| Gene Symbol: Gm12289 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK076620.1 | - | 502 | |||
| BG075615.2 | - | 1 | 437 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB614763.1 | - | 2 | 502 | Chr. 11 - 64012615 - 64013640 [+] on Build GRCm38 | |

Gm12289
Genomic Map