siRNA Details
Gene Symbol: Gm12289 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK076620.1 | - | 394 | |||
BG075615.2 | - | 1 | 329 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB614763.1 | - | 2 | 394 | Chr. 11 - 64012615 - 64013640 [+] on Build GRCm38 |
