siRNA Details
| Gene Symbol: Atxn7l3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK051537.1 | - | 1736 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Atxn7l3b| Gene Symbol: Atxn7l3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK051537.1 | - | 1736 | |||