siRNA Details
| Gene Symbol: Atxn7l3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK037008.1 | - | 1950 | |||
| AK051537.1 | - | 2974 | ||||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB458124.2 | - | 1 | 591 | Chr. 10 - 112923872 - 112924525 [-] on Build GRCm38 | |

Atxn7l3b
Genomic Map