siRNA Details
| Gene Symbol: 4930419G24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040595.1 | 5 | 456 | |||
| GenBank mRNA | AK015168.1 | - | 5 | 457 | ||
| AK029638.1 | - | 5 | 457 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY714714.1 | - | 5 | 457 | Chr. 3 - 33024777 - 33088488 [+] on Build GRCm38 | |

4930419G24Rik
Genomic Map