siRNA Details
| Gene Symbol: 4930419G24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040595.1 | 8 | 1018 | |||
| GenBank mRNA | AK015168.1 | - | 8 | 1019 | ||
| AK029638.1 | - | 8 | 1019 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930419G24Rik| Gene Symbol: 4930419G24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040595.1 | 8 | 1018 | |||
| GenBank mRNA | AK015168.1 | - | 8 | 1019 | ||
| AK029638.1 | - | 8 | 1019 | |||