siRNA Details
| Gene Symbol: 4833411I10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK014679.1 | - | 1 | 492 | ||
| AK138958.1 | - | 1 | 1073 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4833411I10Rik| Gene Symbol: 4833411I10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK014679.1 | - | 1 | 492 | ||
| AK138958.1 | - | 1 | 1073 | |||