siRNA Details
| Gene Symbol: Glis3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK014672.1 | - | 489 | |||
| AK040264.1 | - | 342 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Glis3| Gene Symbol: Glis3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK014672.1 | - | 489 | |||
| AK040264.1 | - | 342 | ||||