siRNA Details
| Gene Symbol: 4833411C07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_102285.1 | 2 | 400 | |||
| GenBank mRNA | AK014677.1 | - | 2 | 311 | ||
| AK050245.1 | - | 2 | 401 | |||
| BC052524.1 | - | 394 | ||||
| BY714173.1 | - | 2 | 311 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4833411C07Rik| Gene Symbol: 4833411C07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_102285.1 | 2 | 400 | |||
| GenBank mRNA | AK014677.1 | - | 2 | 311 | ||
| AK050245.1 | - | 2 | 401 | |||
| BC052524.1 | - | 394 | ||||
| BY714173.1 | - | 2 | 311 | |||