siRNA Details
| Gene Symbol: 4833411C07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_102285.1 | 1, 2 | 237 | |||
| GenBank mRNA | AK014677.1 | - | 1, 2 | 148 | ||
| AK050245.1 | - | 1, 2 | 238 | |||
| BC052524.1 | - | 231 | ||||
| BY714173.1 | - | 1, 2 | 148 | |||

4833411C07Rik
Genomic Map