siRNA Details
Gene Symbol: Kcnj9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC038600.1 | - | 1 | 1297 | ||
BC048090.1 | - | 1 | 686 | |||
BC069221.1 | - | 1 | 221 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Kcnj9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC038600.1 | - | 1 | 1297 | ||
BC048090.1 | - | 1 | 686 | |||
BC069221.1 | - | 1 | 221 |