siRNA Details
| Gene Symbol: Gm29856 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK082514.1 | - | 2 | 344 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB651099.1 | - | 2 | 341 | Chr. 1 - 177458969 - 177460617 [+] on Build GRCm38 | |

Gm29856
Genomic Map