siRNA Details
| Gene Symbol: Gm41829 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK089320.1 | - | 1 | 1572 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY764273.1 | - | 1 | 58 | Chr. 19 - 29239448 - 29240146 [-] on Build GRCm38 | |

Gm41829
Genomic Map