siRNA Details
| Gene Symbol: G3bp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK080910.1 | - | 979 | |||
| AK083741.1 | - | 1027 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

G3bp1| Gene Symbol: G3bp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK080910.1 | - | 979 | |||
| AK083741.1 | - | 1027 | ||||