siRNA Details
| Gene Symbol: Snhg5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040721.1 | 5 | 867 | |||
| GenBank mRNA | AK012825.1 | - | 5 | 859 | ||
| BC032970.1 | - | 5 | 842 | |||
| KF876690.1 | - | 5 | 861 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Snhg5| Gene Symbol: Snhg5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040721.1 | 5 | 867 | |||
| GenBank mRNA | AK012825.1 | - | 5 | 859 | ||
| BC032970.1 | - | 5 | 842 | |||
| KF876690.1 | - | 5 | 861 | |||