siRNA Details
| Gene Symbol: Cspp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK009423.1 | - | 1 | 866 | ||
| AK134458.1 | - | 1 | 3014 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cspp1| Gene Symbol: Cspp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK009423.1 | - | 1 | 866 | ||
| AK134458.1 | - | 1 | 3014 | |||