siRNA Details
| Gene Symbol: Gm7759 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK135850.1 | - | 2 | 194 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY719791.1 | - | 2 | 194 | Chr. 1 - 22898387 - 22904303 [+] on Build GRCm38 | |

Gm7759
Genomic Map