siRNA Details
| Gene Symbol: Wdfy2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK031001.1 | - | 1 | 2470 | ||
| AK164078.1 | - | 1 | 2151 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Wdfy2| Gene Symbol: Wdfy2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK031001.1 | - | 1 | 2470 | ||
| AK164078.1 | - | 1 | 2151 | |||