siRNA Details
| Gene Symbol: 4930486F22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038024.1 | 3 | 516 | |||
| GenBank mRNA | AK019657.1 | - | 3 | 512 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY715184.1 | - | 3 | 512 | Chr. 10 - 86095859 - 86113586 [+] on Build GRCm38 | |

4930486F22Rik
Genomic Map