siRNA Details
| Gene Symbol: 4930486F22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038024.1 | 3 | 279 | |||
| GenBank mRNA | AK019657.1 | - | 3 | 275 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930486F22Rik| Gene Symbol: 4930486F22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038024.1 | 3 | 279 | |||
| GenBank mRNA | AK019657.1 | - | 3 | 275 | ||