siRNA Details
| Gene Symbol: 1810019N24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK019651.1 | - | 1 | 68 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY715158.1 | - | 1 | 68 | Chr. 7 - 12949690 - 12950066 [-] on Build GRCm38 | |

1810019N24Rik
Genomic Map