siRNA Details
| Gene Symbol: 4930405N21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK015101.1 | - | 1 | 575 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY714634.1 | - | 1 | 573 | Chr. 5 - 113320825 - 113321704 [-] on Build GRCm38 | |

4930405N21Rik
Genomic Map