siRNA Details
| Gene Symbol: 2700054A10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK137633.1 | - | 209 | |||
| BC068229.1 | - | 54 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

2700054A10Rik| Gene Symbol: 2700054A10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK137633.1 | - | 209 | |||
| BC068229.1 | - | 54 | ||||