siRNA Details
| Gene Symbol: 4930553M12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK016108.1 | - | 1 | 469 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY715712.1 | - | 469 | Chr. - - [-] on Build | ||

4930553M12Rik
Genomic Map