siRNA Details
| Gene Symbol: 4930553J12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK016107.1 | - | 493 | |||
| BC062172.1 | - | 1 | 367 | |||
| BC080822.1 | - | 1 | 355 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY715711.1 | - | 4 | 493 | Chr. 16 - 88798892 - 88820671 [+] on Build GRCm38 | |

4930553J12Rik
Genomic Map