siRNA Details
Gene Symbol: 4930553E22Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040567.1 | 3 | 344 | |||
GenBank mRNA | AK016102.1 | - | 3 | 345 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 4930553E22Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040567.1 | 3 | 344 | |||
GenBank mRNA | AK016102.1 | - | 3 | 345 |