siRNA Details
| Gene Symbol: Luc7l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK042182.1 | - | 1 | 797 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Luc7l2| Gene Symbol: Luc7l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK042182.1 | - | 1 | 797 | ||