siRNA Details
| Gene Symbol: 1810019D21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040344.1 | 3 | 543 | |||
| NR_040345.1 | 3 | 543 | ||||
| NR_040346.1 | 2 | 274 | ||||
| GenBank mRNA | AK007545.1 | - | 1 | 151 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1810019D21Rik| Gene Symbol: 1810019D21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040344.1 | 3 | 543 | |||
| NR_040345.1 | 3 | 543 | ||||
| NR_040346.1 | 2 | 274 | ||||
| GenBank mRNA | AK007545.1 | - | 1 | 151 | ||